Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GPR15 Antibody (367902), Biotin, Novus Biologicals™
SDP

Catalog No. NB371840 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB371840 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB371840 Supplier Novus Biologicals Supplier No. FAB3654B
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

GPR15 Monoclonal antibody specifically detects GPR15 in Human samples. It is validated for Flow Cytometry, CyTOF-reported

Specifications

Antigen GPR15
Applications Flow Cytometry, CyTOF
Classification Monoclonal
Clone 367902
Conjugate Biotin
Dilution Flow Cytometry, CyTOF-reported
Formulation PBS
Gene Alias BOB, Brother of Bonzo, G protein-coupled receptor 15, G-protein coupled receptor 15, MGC126828, MGC126830
Host Species Mouse
Immunogen NS0 mouse myeloma cell line transfected with human GPR15, Met1-Leu360, Accession # P49685
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline GPCR
Primary or Secondary Primary
Gene ID (Entrez) 2838
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.