Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GPR15 Antibody (367902), DyLight 350, Novus Biologicals™
SDP

Catalog No. NB371838 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB371838 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB371838 Supplier Novus Biologicals Supplier No. FAB3654D
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

GPR15 Monoclonal specifically detects GPR15 in Human samples. It is validated for CyTOF, Flow Cytometry.

Specifications

Antigen GPR15
Applications CyTOF, Flow Cytometry
Classification Monoclonal
Clone 367902
Conjugate DyLight 350
Dilution Flow Cytometry, CyTOF-reported
Formulation 50mM Sodium Borate
Gene Alias BOB, Brother of Bonzo, G protein-coupled receptor 15, G-protein coupled receptor 15, MGC126828, MGC126830
Host Species Mouse
Immunogen NS0 mouse myeloma cell line transfected with human GPR15, Met1-Leu360, Accession # P49685
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline GPCR
Primary or Secondary Primary
Gene ID (Entrez) 2838
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.