Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GPR150 Rabbit anti-Human, Alexa Fluor 750, Polyclonal, Novus Biologicals™
SDP

Catalog No. NB145695 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB145695 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB145695 Supplier Novus Biologicals Supplier No. NBP306009AF750

Rabbit Polyclonal Antibody

GPR150 Polyclonal specifically detects GPR150 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen GPR150
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Alexa Fluor 750
Dilution Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias G protein-coupled receptor 150, PGR11, probable G-protein coupled receptor 150, seven transmembrane helix receptor
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human GPR150 fragment.
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline GPCR
Primary or Secondary Primary
Gene ID (Entrez) 285601
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.