Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GPR153 Antibody - BSA Free, Novus Biologicals™
SDP

Catalog No. NLS2032 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.05 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NLS2032 0.05 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NLS2032 Supplier Novus Biologicals Supplier No. NLS2032
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

GPR153 Polyclonal specifically detects GPR153 in Human, Mouse, Rat, Porcine, Bovine, Canine, Hamster, Monkey, Rabbit samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen GPR153
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Immunohistochemistry, Immunohistochemistry-Paraffin 2 - 12 ug/ml
Formulation PBS with 0.1% Sodium Azide
Gene Alias DKFZp762B2210, G protein-coupled receptor 153, PGR1G-protein coupled receptor PGR1, probable G-protein coupled receptor 153
Gene Symbols GPR153
Host Species Rabbit
Immunogen Synthetic 17 amino acid peptide from 2nd extracellular domain of human GPR153.
Purification Method Affinity Purified
Quantity 0.05 mL
Regulatory Status RUO
Research Discipline GPCR
Primary or Secondary Primary
Gene ID (Entrez) 387509
Test Specificity G Protein-Coupled Receptor LS19072
Target Species Human, Mouse, Rat, Pig, Bovine, Canine, Hamster, Monkey, Rabbit
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.