Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GPR154 Antibody - BSA Free, Novus Biologicals™
SDP

Catalog No. NBP144315 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.05 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP144315 0.05 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP144315 Supplier Novus Biologicals Supplier No. NBP144315
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

GPR154 Polyclonal specifically detects GPR154 in Human, Mouse, Rat, Hamster, Monkey samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen GPR154
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Immunohistochemistry, Immunohistochemistry-Paraffin 5 ug/ml
Formulation PBS with 0.1% Sodium Azide
Gene Alias G protein-coupled receptor 154, G protein-coupled receptor for asthma susceptibility, GPR154NPSR, GPRAASRT2, G-protein coupled receptor 154, G-protein coupled receptor for asthma susceptibility, G-protein coupled receptor PGR14, neuropeptide S receptor, neuropeptide S receptor 1, PGR14VRR1, vasopressin receptor-related receptor 1
Gene Symbols NPSR1
Host Species Rabbit
Immunogen Synthetic 18 amino acid peptide from 2nd extracellular domain of human GPR154.
Purification Method Affinity Purified
Quantity 0.05 mg
Regulatory Status RUO
Research Discipline GPCR
Primary or Secondary Primary
Gene ID (Entrez) 387129
Test Specificity This product is specific for G-Coupled GPR154. Unless other species reactivities are specified there is no testing data currently available.
Target Species Human, Mouse, Rat, Hamster, Monkey
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.