Learn More
Description
Specifications
Specifications
| Antigen | GPR154 |
| Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Dilution | Immunohistochemistry 1:500 - 1:1000, Immunohistochemistry-Paraffin 1:500 - 1:1000 |
| Formulation | PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide |
| Gene Accession No. | Q6W5P4 |
| Gene Alias | G protein-coupled receptor 154, G protein-coupled receptor for asthma susceptibility, GPR154NPSR, GPRAASRT2, G-protein coupled receptor 154, G-protein coupled receptor for asthma susceptibility, G-protein coupled receptor PGR14, neuropeptide S receptor, neuropeptide S receptor 1, PGR14VRR1, vasopressin receptor-related receptor 1 |
| Gene Symbols | NPSR1 |
| Host Species | Rabbit |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
