Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GPR155 Antibody, Alexa Fluor™ 532, Novus Biologicals™
SDP

Catalog No. NP1062A32 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NP1062A32 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NP1062A32 Supplier Novus Biologicals Supplier No. NBP150625AF532
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

GPR155 Polyclonal specifically detects GPR155 in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen GPR155
Applications Western Blot, Immunocytochemistry, Immunofluorescence
Classification Polyclonal
Conjugate Alexa Fluor 532
Dilution Western Blot, Immunocytochemistry/Immunofluorescence
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias DEP.7, DEPDC3, FLJ31819, FLJ39346, G protein-coupled receptor 155, integral membrane protein GPR155, PGR22G-protein coupled receptor PGR22
Gene Symbols GPR155
Host Species Rabbit
Immunogen A synthetic peptide made to an internal portion of the human GPR155 protein (between residues 500-600) [UniProt Q7Z3F1]
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 151556
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.