Learn More
Description
Specifications
Specifications
| Antigen | GPR155 |
| Applications | Western Blot, Immunocytochemistry, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | DyLight 350 |
| Dilution | Western Blot, Immunocytochemistry/Immunofluorescence |
| Formulation | 50 mM sodium borate with 0.05% sodium azide |
| Gene Alias | DEP.7, DEPDC3, FLJ31819, FLJ39346, G protein-coupled receptor 155, integral membrane protein GPR155, PGR22G-protein coupled receptor PGR22 |
| Gene Symbols | GPR155 |
| Host Species | Rabbit |
| Immunogen | A synthetic peptide made to an internal portion of the human GPR155 protein (between residues 800-900) [UniProt Q7Z3F1] |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
