Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GPR158 Antibody (1027651) [Biotin], Novus Biologicals™
SDP

Catalog No. NB402053 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB402053 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB402053 Supplier Novus Biologicals Supplier No. FAB102861B
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

GPR158 Monoclonal antibody specifically detects GPR158 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry

Specifications

Antigen GPR158
Applications Flow Cytometry, Immunocytochemistry
Classification Monoclonal
Clone 1027651
Conjugate Biotin
Dilution Flow Cytometry, Immunocytochemistry
Formulation PBS
Gene Alias G protein-coupled receptor 158, KIAA1136FLJ37801, probable G-protein coupled receptor 158, RP11-59G22.1
Host Species Mouse
Immunogen Human embryonic kidney cell HEK293-derived human GPR158 protein, Ala24-Gln411, Accession # Q5T848
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline GPCR
Primary or Secondary Primary
Gene ID (Entrez) 57512
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.