Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GPR15L Antibody (1020904) [DyLight 405], Novus Biologicals™
SDP

Catalog No. NB401584 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB401584 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB401584 Supplier Novus Biologicals Supplier No. FAB104161E
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

GPR15L Monoclonal antibody specifically detects GPR15L in Human samples. It is validated for ELISA, Flow Cytometry (Intracellular Staining), CyTOF

Specifications

Antigen GPR15L
Applications ELISA, Flow Cytometry (Intracellular Staining), CyTOF
Classification Monoclonal
Clone 1020904
Conjugate DyLight 405
Dilution ELISA, Intracellular Staining by Flow Cytometry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias C10orf99 chromosome 10 open reading frame 99, UNQ1833
Host Species Mouse
Immunogen E. coli-derived human C10orf99
Purification Method Protein A or G purified from cell culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 387695
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.