Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GPR18 Antibody - BSA Free, Novus Biologicals™
SDP

Catalog No. NBP224918SS Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP224918SS 0.025 mg
NBP224918 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP224918SS Supplier Novus Biologicals Supplier No. NBP224918SS
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 3 publications

GPR18 Polyclonal specifically detects GPR18 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen GPR18
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 2 ug/ml, Flow Cytometry reported in scientific literature (PMID 27256622), Immunohistochemistry 1:200, Immunocytochemistry/Immunofluorescence 2 - 10 ug/ml. Use reported in scientific literature (PMID 30894369), Immunohistochemistry-Paraffin 1:200
Formulation PBS with 0.05% Sodium Azide
Gene Alias G protein-coupled receptor 18, GPCRW, G-protein coupled receptor 18, NAGly receptor, N-arachidonyl glycine receptor
Gene Symbols GPR18
Host Species Rabbit
Immunogen A portion of amino acids 1-50 of human GPR18 was used as the immunogen.
Purification Method Affinity Purified
Quantity 0.025 mg
Regulatory Status RUO
Research Discipline GPCR
Primary or Secondary Primary
Gene ID (Entrez) 2841
Target Species Human, Mouse, Rat
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.