Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GPR18 Antibody, PE, Novus Biologicals™
SDP

Catalog No. NB141980 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB141980 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB141980 Supplier Novus Biologicals Supplier No. NBP224918PE
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

GPR18 Polyclonal specifically detects GPR18 in Human, Mouse, Rat samples. It is validated for Flow Cytometry.

Specifications

Antigen GPR18
Applications Flow Cytometry
Classification Polyclonal
Conjugate PE
Dilution Flow Cytometry
Formulation PBS
Gene Alias G protein-coupled receptor 18, GPCRW, G-protein coupled receptor 18, NAGly receptor, N-arachidonyl glycine receptor
Host Species Rabbit
Immunogen Partial synthetic peptide made to an N-terminal portion of the human GPR18 protein (between amino acids 1-50) [UniProt Q14330]
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline GPCR
Primary or Secondary Primary
Gene ID (Entrez) 2841
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.