Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GPR22 Antibody (455108), Alexa Fluor™ 350, Novus Biologicals™
SDP

Catalog No. NB296260 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB296260 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB296260 Supplier Novus Biologicals Supplier No. FAB4500U

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Mouse Monoclonal Antibody

GPR22 Monoclonal antibody specifically detects GPR22 in Human samples. It is validated for Flow Cytometry, CyTOF

Specifications

Antigen GPR22
Applications Flow Cytometry, CyTOF
Classification Monoclonal
Clone 455108
Conjugate Alexa Fluor 350
Dilution Intracellular Staining by Flow Cytometry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias G protein-coupled receptor 22, MGC129847, probable G-protein coupled receptor 22
Host Species Mouse
Immunogen HEK293 human embryonic kidney cell line transfected with human GPR22, Met1-Asp433, Accession # Q99680
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline GPCR
Primary or Secondary Primary
Gene ID (Entrez) 2845
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.