Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GPR27 Antibody, Novus Biologicals™
SDP

Catalog No. NB158129 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB158129 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB158129 Supplier Novus Biologicals Supplier No. NBP314357
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

GPR27 Polyclonal antibody specifically detects GPR27 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)

Specifications

Antigen GPR27
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation PBS
Gene Alias G protein-coupled receptor 27, probable G-protein coupled receptor 27, SREB1Super conserved receptor expressed in brain 1
Host Species Rabbit
Immunogen Synthetic 19 amino acid peptide from 3rd cytoplasmic domain of human GPR27.
Purification Method Immunogen affinity purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline GPCR
Primary or Secondary Primary
Gene ID (Entrez) 2850
Target Species Human
Content And Storage Keep as concentrated solution. Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Product Type Antibody
Form Purified
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.