Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Novus Biologicals™ Collagen VI alpha 3 Recombinant Protein Antigen
Shop All R&D Systems Products

Click to view available options
Quantity:
0.1 mL
Description
A recombinant protein antigen with a N-terminal His6-ABP tag corresponding Collagen VI alpha 3. The Collagen VI alpha 3 Recombinant Protein Antigen is derived from E. coli. The Collagen VI alpha 3 Recombinant Protein Antigen has been validated for the following applications: Antibody Competition.
Specifications
Specifications
| Gene ID (Entrez) | 1293 |
| Purity | >80%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie™ Blue stain |
| Common Name | Collagen VI alpha 3 |
| Content And Storage | Store at −20°C. Avoid freeze-thaw cycles. |
| Formulation | PBS and 1M Urea, pH 7.4. |
| For Use With (Application) | This Recombinant Antigen is Only Intended to be Used as a Blocking Agent to Confirm Antibody Specificity with The Corresponding Antibody, Catalog Number NBP2-49678. It is Purified by IMAC Chromatography, and The Expected Concentration is Greater Than 0.5 mg/ml |
| Gene Alias | Collagen Alpha-3(VI) Chain, Collagen VI, Alpha-3 Polypeptide, collagen, type VI, alpha 3 |
| Quantity | 0.1 mL |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction