Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GPR35 Monoclonal Antibody (C3), Invitrogen™
GREENER_CHOICE

Mouse Monoclonal Antibody

$255.00 - $547.00

Specifications

Antigen GPR35
Clone C3
Concentration 1 mg/mL
Applications Western Blot
Classification Monoclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
PIMA543796
View Documents
Invitrogen™
MA543796
20 μL
Each for $255.00
Only null left
Add to Cart
 
PIMA543795
GREENER_CHOICE
View Documents
Invitrogen™
MA543795
100 μL
Each for $547.00
Only null left
Add to Cart
 
Description

Description

Acts as a receptor for kynurenic acid, an intermediate in the tryptophan metabolic pathway. The activity of this receptor is mediated by G-proteins that elicit calcium mobilization and inositol phosphate production through G(qi/o) proteins. [UniProt]
Specifications

Specifications

GPR35
1 mg/mL
Monoclonal
Liquid
RUO
PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4
G protein-coupled receptor 35; Gpr35; G-protein coupled receptor 35; KPG_007; KYNA receptor; kynurenic acid receptor
GPR35
IgG1 κ
Protein A/G
Antibody
C3
Western Blot
Unconjugated
Mouse
Human
Q9HC97
2859
Arg240-Ala309 (Accession Q9HC97), with N-terminal His and GST Tag.
Primary
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
GPR35
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.