Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GPR37 Antibody (436923), Allophycocyanin, Novus Biologicals™
SDP

Catalog No. NB370071 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB370071 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB370071 Supplier Novus Biologicals Supplier No. FAB4450A
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

GPR37 Monoclonal specifically detects GPR37 in Human samples. It is validated for CyTOF, Flow Cytometry (Intracellular Staining).

Specifications

Antigen GPR37
Applications CyTOF, Flow Cytometry (Intracellular Staining)
Classification Monoclonal
Clone 436923
Conjugate APC
Dilution Intracellular Staining by Flow Cytometry, CyTOF-ready
Formulation PBS
Gene Alias EDNRBL, Endothelin B receptor-like protein 1, G protein-coupled receptor 37 (endothelin receptor type B-like), hET(B)R-LP, PAELRETBR-LP-1, Parkin-associated endothelin receptor-like receptor, probable G-protein coupled receptor 37
Host Species Mouse
Immunogen NS0 mouse myeloma cell line transfected with human GPR37, Ala27-Cys613, Accession # O15354
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline GPCR
Primary or Secondary Primary
Gene ID (Entrez) 2861
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.