Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GPR37L1 Antibody (444532), DyLight 488, Novus Biologicals™
SDP

Catalog No. NB370079 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB370079 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB370079 Supplier Novus Biologicals Supplier No. FAB4449K
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

GPR37L1 Monoclonal specifically detects GPR37L1 in Human samples. It is validated for CyTOF, Flow Cytometry (Intracellular Staining).

Specifications

Antigen GPR37L1
Applications CyTOF, Flow Cytometry (Intracellular Staining)
Classification Monoclonal
Clone 444532
Conjugate DyLight 488
Dilution Intracellular Staining by Flow Cytometry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias endothelin B receptor-like protein 2, ET(B)R-LP-2, ETBRLP2, ETBR-LP-2endothelin type b receptor-like protein 2, G protein-coupled receptor 37 like 1, G-protein coupled receptor 37 like 1, G-protein coupled receptor 37-like 1
Host Species Mouse
Immunogen NS0 mouse myeloma cell line transfected with human GPR37L1, Accession # O60883
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline GPCR, Neuroscience, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 9283
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.