Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GPR39 Antibody (189702) [Janelia Fluor™ 646], Novus Biologicals™
SDP

Catalog No. NB372448 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB372448 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB372448 Supplier Novus Biologicals Supplier No. FAB3348J
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

GPR39 Monoclonal antibody specifically detects GPR39 in Human samples. It is validated for Flow Cytometry, CyTOF

Specifications

Antigen GPR39
Applications Flow Cytometry, CyTOF
Classification Monoclonal
Clone 189702
Conjugate Janelia Fluor 646
Dilution Flow Cytometry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias G protein-coupled receptor 39, G-protein coupled receptor 39, MGC149541
Host Species Mouse
Immunogen NS0 mouse myeloma cell line transfected with human GPR39, Met1-Val453, Accession # BAG37409
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline GPCR, Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 2863
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.