Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GPR39 Antibody - BSA Free, Novus Biologicals™
SDP

Catalog No. NLS142 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.05 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NLS142 0.05 mL
NLS142SS 0.025 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NLS142 Supplier Novus Biologicals Supplier No. NLS142
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 3 publications

GPR39 Polyclonal specifically detects GPR39 in Human, Mouse samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen GPR39
Applications Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1.07 mg/mL
Conjugate Unconjugated
Dilution Immunohistochemistry 1:50, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin 1:50
Formulation PBS and 30% Glycerol with 0.05% Sodium Azide
Gene Accession No. O43194, Q5U431
Gene Alias G protein-coupled receptor 39, G-protein coupled receptor 39, MGC149541
Gene Symbols GPR39
Host Species Rabbit
Immunogen A synthetic peptide made to an internal region of the human GPR39 protein (within residues 400-500). [Swiss-Prot O43194]
Purification Method Affinity Purified
Quantity 0.05 mL
Regulatory Status RUO
Research Discipline GPCR, Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 2863
Target Species Human, Mouse
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.