Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GPR50 Antibody (461129), Janelia Fluor™ 549, Novus Biologicals™
SDP

Catalog No. NB369489 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB369489 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB369489 Supplier Novus Biologicals Supplier No. FAB4645I

Mouse Monoclonal Antibody

GPR50 Monoclonal antibody specifically detects GPR50 in Human samples. It is validated for Flow Cytometry (Intracellular Staining), Immunocytochemistry, CyTOF

Specifications

Antigen GPR50
Applications Flow Cytometry (Intracellular Staining), Immunocytochemistry, CyTOF
Classification Monoclonal
Clone 461129
Conjugate Janelia Fluor 549
Dilution Intracellular Staining by Flow Cytometry, Immunocytochemistry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias G protein-coupled receptor 50H9Mel1c, melatonin-related receptor, MGC125342
Host Species Mouse
Immunogen NS0 mouse myeloma cell line transfected with human GPR50, Met1-Val617, Accession # Q13585
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline GPCR, Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 9248
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.