Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GPR52 Antibody (582671) [DyLight 755], Novus Biologicals™
SDP

Catalog No. NB401650 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB401650 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB401650 Supplier Novus Biologicals Supplier No. FAB10357Z
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

GPR52 Monoclonal antibody specifically detects GPR52 in Human samples. It is validated for Flow Cytometry, CyTOF

Specifications

Antigen GPR52
Applications Flow Cytometry, CyTOF
Classification Monoclonal
Clone 582671
Conjugate DyLight 755
Dilution Flow Cytometry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias G protein-coupled receptor 52, MGC111751, probable G-protein coupled receptor 52
Host Species Mouse
Immunogen NS0 mouse myeloma cell line transfected with human GPR52, Met1-Ile361, Accession # Q9Y2T5
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline GPCR
Primary or Secondary Primary
Gene ID (Entrez) 9293
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.