Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ GPR52 Comp-43
GREENER_CHOICE

Catalog No. p-7222996
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

GPR52 receptor antagonist

GPR52 Comp-43 is a GPR52 receptor antagonist (IC50 = 0.63 μM). The use of GPR52 Comp-43 in animal models of Huntington's Disease leads to degradation and reduction of soluble and aggregated mutant huntingtin protein in the striatum. It also reduces neuronal loss, promotes neuronal survival and improves motor functions in vivo. GPR52 Comp-43 is active in vitro and brain penetrant.

Specifications

Chemical Name or Material (2 E)-5-Phenyl-1-(2-thienyl)-2-penten-1-one
CAS 1239987-91-7
Quantity 10 mg
Target Orphan 7-TM Receptor Antagonists
Molecular Formula C15H14OS
Purity 0.98
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.