Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GPR56 Antibody (776123), PE/Cy7, Novus Biologicals™
SDP

Catalog No. NB312353 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB312353 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB312353 Supplier Novus Biologicals Supplier No. FAB4636PECY7
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

GPR56 Monoclonal specifically detects GPR56 in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen GPR56
Applications Flow Cytometry
Classification Monoclonal
Clone 776123
Conjugate PE-Cyanine7
Dilution Flow Cytometry
Formulation PBS
Gene Alias BFPP, G protein-coupled receptor 56,7-transmembrane protein with no EGF-like N-terminal domains-1, Protein TM7XN1, TM7LN4G-protein coupled receptor 56, TM7XN1DKFZp781L1398
Host Species Mouse
Immunogen Chinese hamster ovary cell line CHO-derived recombinant human GPR56, Met1-Val342, Accession # Q9Y653
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline GPCR, Growth and Development, Neuronal Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 9289
Target Species Human
Content And Storage Store at 4C in the dark. Do not freeze.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.