Learn More
Description
Specifications
Specifications
| Antigen | GPR64 |
| Applications | Flow Cytometry, CyTOF |
| Classification | Monoclonal |
| Clone | 206420 |
| Conjugate | DyLight 550 |
| Dilution | Flow Cytometry, CyTOF-ready |
| Formulation | 50mM Sodium Borate |
| Gene Alias | EDDM6, epididymis-specific (seven transmembrane family), G protein-coupled receptor 64, MGC104454, MGC138738, TM7LN2 |
| Host Species | Mouse |
| Immunogen | Chinese hamster ovary cell line CHO-derived recombinant human GPR64, Leu38-Asn64, Glu68-Thr553, Accession # NP_001073328 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
