Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GPR64 Antibody (864238), DyLight 594, Novus Biologicals™
SDP

Catalog No. NB373800 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB373800 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB373800 Supplier Novus Biologicals Supplier No. FAB79771M
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

GPR64 Monoclonal antibody specifically detects GPR64 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, CyTOF

Specifications

Antigen GPR64
Applications Flow Cytometry, Immunohistochemistry, CyTOF
Classification Monoclonal
Clone 864238
Conjugate DyLight 594
Dilution Flow Cytometry, Immunohistochemistry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias EDDM6, epididymis-specific (seven transmembrane family), G protein-coupled receptor 64, MGC104454, MGC138738, TM7LN2
Host Species Mouse
Immunogen Chinese hamster ovary cell line CHO-derived recombinant human GPR64, Leu38-Asn64, Glu68-Thr553, Accession # NP_001073328
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline GPCR
Primary or Secondary Primary
Gene ID (Entrez) 10149
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.