Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GPR74/NPFFR2 Antibody, Allophycocyanin, Novus Biologicals™
SDP

Catalog No. NB360732 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB360732 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB360732 Supplier Novus Biologicals Supplier No. NBP224878APC
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

GPR74/NPFFR2 Polyclonal antibody specifically detects GPR74/NPFFR2 in Human samples. It is validated for Western Blot

Specifications

Antigen GPR74/NPFFR2
Applications Western Blot
Classification Polyclonal
Conjugate APC
Dilution Western Blot
Formulation PBS
Gene Alias GPR74G protein-coupled receptor 74, G-protein coupled receptor 74, neuropeptide FF 2, neuropeptide FF receptor 2, Neuropeptide G-protein coupled receptor, NPFF2G-protein coupled receptor HLWAR77, NPGPRHLWAR77
Host Species Rabbit
Immunogen Reacts with the human NPFF2 protein.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline GPCR, Neuronal Cell Markers, Neuroscience, Neurotransmission
Primary or Secondary Primary
Gene ID (Entrez) 10886
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.