Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GPR74/NPFFR2 Antibody - BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB300169SS
Description
GPR74/NPFFR2 Polyclonal specifically detects GPR74/NPFFR2 in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
GPR74/NPFFR2 | |
Polyclonal | |
Unconjugated | |
PBS with 0.02% Sodium Azide | |
NPFFR2 | |
N terminal sequence MNEKWDTNSSENWHPI and the C terminal sequence ELVMEELKETTNSSEI of the human NPFF2 protein. | |
0.025 mg | |
GPCR, Neuronal Cell Markers, Neuroscience, Neurotransmission | |
10886 | |
Human | |
IgG |
Immunofluorescence, Immunocytochemistry, Western Blot | |
1 mg/mL | |
Western Blot 1:500 -1:1000, Immunocytochemistry/Immunofluorescence | |
GPR74G protein-coupled receptor 74, G-protein coupled receptor 74, neuropeptide FF 2, neuropeptide FF receptor 2, Neuropeptide G-protein coupled receptor, NPFF2G-protein coupled receptor HLWAR77, NPGPRHLWAR77 | |
Rabbit | |
Affinity Purified | |
RUO | |
Primary | |
Reacts with the N terminal sequence MNEKWDTNSSENWHPI and the C terminal sequence ELVMEELKETTNSSEI of the human NPFF2 protein. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction