Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GPR74/NPFFR2 Antibody - BSA Free, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

$206.50 - $499.50

Specifications

Antigen GPR74/NPFFR2
Concentration 1 mg/mL
Dilution Western Blot 1:500 -1:1000, Immunocytochemistry/Immunofluorescence
Applications Immunofluorescence, Immunocytochemistry, Western Blot
Classification Polyclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB018148
SDP
View Documents
Novus Biologicals
NB300169SS
0.025 mg
Each for $206.50
Only null left
Add to Cart
 
NB300169
SDP
View Documents
Novus Biologicals
NB300169
0.1 mg
Each for $499.50
Only null left
Add to Cart
 
Description

Description

GPR74/NPFFR2 Polyclonal specifically detects GPR74/NPFFR2 in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
Specifications

Specifications

GPR74/NPFFR2
Western Blot 1:500 -1:1000, Immunocytochemistry/Immunofluorescence
Polyclonal
Rabbit
GPCR, Neuronal Cell Markers, Neuroscience, Neurotransmission
PBS with 0.02% Sodium Azide
10886
N terminal sequence MNEKWDTNSSENWHPI and the C terminal sequence ELVMEELKETTNSSEI of the human NPFF2 protein.
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
1 mg/mL
Immunofluorescence, Immunocytochemistry, Western Blot
Unconjugated
RUO
Human
GPR74G protein-coupled receptor 74, G-protein coupled receptor 74, neuropeptide FF 2, neuropeptide FF receptor 2, Neuropeptide G-protein coupled receptor, NPFF2G-protein coupled receptor HLWAR77, NPGPRHLWAR77
NPFFR2
IgG
Affinity Purified
Reacts with the N terminal sequence MNEKWDTNSSENWHPI and the C terminal sequence ELVMEELKETTNSSEI of the human NPFF2 protein.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.