Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GPR83 Antibody (566133), Allophycocyanin, Novus Biologicals™
SDP

Catalog No. NB305409 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB305409 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB305409 Supplier Novus Biologicals Supplier No. FAB10943A
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

GPR83 Monoclonal antibody specifically detects GPR83 in Human samples. It is validated for Flow Cytometry

Specifications

Antigen GPR83
Applications Flow Cytometry
Classification Monoclonal
Clone 566133
Conjugate APC
Dilution Flow Cytometry
Formulation PBS
Gene Alias G protein-coupled receptor 72, G protein-coupled receptor 83, GIR, GPR72glucocorticoid induced receptor, G-protein coupled receptor 72, KIAA1540, probable G-protein coupled receptor 83
Host Species Mouse
Immunogen NS0 mouse myeloma cell line transfected with human GPR83, Accession # Q9NYM4
Purification Method Protein A or G purified from cell culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline GPCR
Primary or Secondary Primary
Gene ID (Entrez) 10888
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.