Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GPR83 Antibody, mFluor Violet 500 SE, Novus Biologicals™
SDP

Catalog No. NB337425 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB337425 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB337425 Supplier Novus Biologicals Supplier No. NBP224515MFV500
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

GPR83 Polyclonal antibody specifically detects GPR83 in Human, Mouse, Rat, Bovine, Canine, Chicken, Primate samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)

Specifications

Antigen GPR83
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
Classification Polyclonal
Conjugate mFluor Violet 500 SE
Dilution Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen
Formulation 50mM Sodium Borate
Gene Alias G protein-coupled receptor 72, G protein-coupled receptor 83, GIR, GPR72glucocorticoid induced receptor, G-protein coupled receptor 72, KIAA1540, probable G-protein coupled receptor 83
Host Species Rabbit
Immunogen A portion of amino acids 200-250 of human GPR83 was used as the immunogen for this antibody.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline GPCR
Primary or Secondary Primary
Gene ID (Entrez) 10888
Target Species Human, Mouse, Rat, Bovine, Canine
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.