Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GPR97 Antibody (2435C) [HRP], Novus Biologicals™
SDP

Catalog No. NB402501 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB402501 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB402501 Supplier Novus Biologicals Supplier No. FAB10097H
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

GPR97 Monoclonal specifically detects GPR97 in Human samples. It is validated for CyTOF, Flow Cytometry, Western Blot.

Specifications

Antigen GPR97
Applications CyTOF, Flow Cytometry, Western Blot
Classification Monoclonal
Clone 2435C
Conjugate HRP
Dilution Western Blot, Flow Cytometry, CyTOF-ready
Formulation PBS
Gene Alias G protein-coupled receptor 97, Pb99, PGR26G-protein coupled receptor PGR26, probable G-protein coupled receptor 97
Host Species Rabbit
Immunogen Human embryonic kidney cell, HEK293-transfected human GPR97, Met1-Leu249, Accession # Q86Y34
Purification Method Protein A or G purified from cell culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline GPCR
Primary or Secondary Primary
Gene ID (Entrez) 222487
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.