Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GPRC5A/RAI3 Antibody (481906), DyLight 755, Novus Biologicals™
SDP

Catalog No. NB368293 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB368293 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB368293 Supplier Novus Biologicals Supplier No. FAB5239Z
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

GPRC5A/RAI3 Monoclonal antibody specifically detects GPRC5A/RAI3 in Human samples. It is validated for Flow Cytometry, Flow Cytometry (Intracellular Staining), CyTOF

Specifications

Antigen GPRC5A/RAI3
Applications Flow Cytometry, Flow Cytometry (Intracellular Staining), CyTOF
Classification Monoclonal
Clone 481906
Conjugate DyLight 755
Dilution Flow Cytometry, Intracellular Staining by Flow Cytometry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias G protein-coupled receptor, family C, group 5, member A, GPCR5A, Orphan G-protein-coupling receptor PEIG-1, RAI3G-protein coupled receptor family C group 5 member A, RAIG1RAIG-1, retinoic acid induced 3, retinoic acid responsive, Retinoic acid-induced gene 1 protein, retinoic acid-induced protein 3
Host Species Mouse
Immunogen NS0 mouse myeloma cell line transfected with human GPRC5A, Met1-Ser357, Accession # Q8NFJ5
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline GPCR
Primary or Secondary Primary
Gene ID (Entrez) 9052
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.