Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GPRC5A/RAI3 Antibody (HL1864) - Azide and BSA Free, Novus Biologicals™
SDP

Catalog No. NB294997 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB294997 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB294997 Supplier Novus Biologicals Supplier No. NBP325620
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

GPRC5A/RAI3 Monoclonal antibody specifically detects GPRC5A/RAI3 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)

Specifications

Antigen GPRC5A/RAI3
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone HL1864
Conjugate Unconjugated
Dilution Western Blot 1:500-1:3000, Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation PBS
Gene Alias G protein-coupled receptor, family C, group 5, member A, GPCR5A, Orphan G-protein-coupling receptor PEIG-1, RAI3G-protein coupled receptor family C group 5 member A, RAIG1RAIG-1, retinoic acid induced 3, retinoic acid responsive, Retinoic acid-induced gene 1 protein, retinoic acid-induced protein 3
Host Species Rabbit
Immunogen Recombinant fragment of human GPRC5A/RAI3.
Purification Method Protein A purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline GPCR
Primary or Secondary Primary
Gene ID (Entrez) 9052
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.