Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GPRC5B Antibody (SR1918), Novus Biologicals™
SDP

Catalog No. NB387881 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB387881 100 μg
NB387880 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB387881 Supplier Novus Biologicals Supplier No. NBP322573100UL
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

GPRC5B Monoclonal antibody specifically detects GPRC5B in Human samples. It is validated for Western Blot

Specifications

Antigen GPRC5B
Applications Western Blot
Classification Monoclonal
Clone SR1918
Conjugate Unconjugated
Dilution Western Blot 1:500-1:2000
Formulation PBS (pH 7.4), 150 mM NaCl, 50% Glycerol
Gene Alias G protein-coupled receptor, family C, group 1, member B, G protein-coupled receptor, family C, group 5, member B, RAIG-2A-69G12.1, RAIG2G-protein coupled receptor family C group 5 member B, retinoic acid responsive gene protein, Retinoic acid-induced gene 2 protein
Host Species Rabbit
Immunogen A synthesized peptide derived from human GPRC5B (Uniprot #: Q9NZH0)
Purification Method Affinity purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline GPCR
Primary or Secondary Primary
Gene ID (Entrez) 51704
Target Species Human
Content And Storage Store at -20°C. Avoid freeze/thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.