Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GPRC5B Antibody (SR1918), Novus Biologicals™

Rabbit Monoclonal Antibody
$302.50 - $413.50
Specifications
Antigen | GPRC5B |
---|---|
Clone | SR1918 |
Dilution | Western Blot 1:500-1:2000 |
Applications | Western Blot |
Classification | Monoclonal |
Description
GPRC5B Monoclonal antibody specifically detects GPRC5B in Human samples. It is validated for Western BlotSpecifications
GPRC5B | |
Western Blot 1:500-1:2000 | |
Monoclonal | |
Purified | |
RUO | |
Human | |
G protein-coupled receptor, family C, group 1, member B, G protein-coupled receptor, family C, group 5, member B, RAIG-2A-69G12.1, RAIG2G-protein coupled receptor family C group 5 member B, retinoic acid responsive gene protein, Retinoic acid-induced gene 2 protein | |
A synthesized peptide derived from human GPRC5B (Uniprot #: Q9NZH0) | |
Primary | |
Store at -20°C. Avoid freeze/thaw cycles. |
SR1918 | |
Western Blot | |
Unconjugated | |
Rabbit | |
GPCR | |
PBS (pH 7.4), 150 mM NaCl, 50% Glycerol | |
51704 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title