Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GPRC5C Antibody (577315) [DyLight 650], Novus Biologicals™
SDP

Catalog No. NB376152 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB376152 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB376152 Supplier Novus Biologicals Supplier No. FAB6594W
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

GPRC5C Monoclonal antibody specifically detects GPRC5C in Human samples. It is validated for Flow Cytometry, CyTOF

Specifications

Antigen GPRC5C
Applications Flow Cytometry, CyTOF
Classification Monoclonal
Clone 577315
Conjugate DyLight 650
Dilution Flow Cytometry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias G protein-coupled receptor, family C, group 5, member C, MGC131820, RAIG-3G-protein coupled receptor family C group 5 member C, RAIG3orphan G-protein coupled receptor, retinoic acid responsive gene protein, Retinoic acid-induced gene 3 protein
Host Species Mouse
Immunogen NS0 mouse myeloma cell line transfected with recombinant human GPRC5C, Accession # Q9NQ84
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline GPCR
Primary or Secondary Primary
Gene ID (Entrez) 55890
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.