Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GPRC5D Antibody (19), mFluor Violet 610 SE, Novus Biologicals™
SDP

Catalog No. NB292786 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB292786 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB292786 Supplier Novus Biologicals Supplier No. NBP326101MFV610
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

GPRC5D Monoclonal antibody specifically detects GPRC5D in Human samples. It is validated for Flow Cytometry

Specifications

Antigen GPRC5D
Applications Flow Cytometry
Classification Monoclonal
Clone 19
Conjugate mFluor Violet 610 SE
Dilution Flow Cytometry
Formulation 50mM Sodium Borate
Gene Alias G protein-coupled receptor, family C, group 5, member D, G-protein coupled receptor family C group 5 member D, MGC129713, MGC129714, orphan G-protein coupled receptor
Host Species Mouse
Immunogen This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human GPRC5D.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline GPCR
Primary or Secondary Primary
Gene ID (Entrez) 55507
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.