Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GPRC5D Antibody (571961R) [DyLight 680], Novus Biologicals™
SDP

Catalog No. NB376710 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB376710 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB376710 Supplier Novus Biologicals Supplier No. FAB6300RY

Mouse Monoclonal Antibody

GPRC5D Monoclonal antibody specifically detects GPRC5D in Human samples. It is validated for Flow Cytometry, CyTOF

Specifications

Antigen GPRC5D
Applications Flow Cytometry, CyTOF
Classification Monoclonal
Clone 571961R
Conjugate DyLight 680
Dilution Flow Cytometry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias G protein-coupled receptor, family C, group 5, member D, G-protein coupled receptor family C group 5 member D, MGC129713, MGC129714, orphan G-protein coupled receptor
Host Species Mouse
Immunogen NS0 mouse myeloma cell line transfected with human GPRC5D, Met1-Val345, Accession # Q9NZD1
Purification Method Protein A or G purified from cell culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline GPCR
Primary or Secondary Primary
Gene ID (Entrez) 55507
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.