Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GPSM2 Antibody, Novus Biologicals™
SDP

Catalog No. NB406713 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
25ul
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB406713 25ul
NBP185231 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB406713 Supplier Novus Biologicals Supplier No. NBP18523125UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

GPSM2 Polyclonal specifically detects GPSM2 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen GPSM2
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 0.4 ug/ml, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1-4 ug/ml, Immunohistochemistry-Paraffin 1:50 - 1:200
Formulation PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide
Gene Accession No. P81274
Gene Alias deafness, autosomal recessive 82, DFNB82, G-protein signaling modulator 2, G-protein signalling modulator 2 (AGS3-like, C. elegans), G-protein-signaling modulator 2, LGNPins, Mosaic protein LGN, PINS
Gene Symbols GPSM2
Host Species Rabbit
Immunogen This antibody was developed against Recombinant Protein corresponding to amino acids:FQSNRMDDQRCCLQEKNCHTASTTTSSTPPKMMLKTSSVPVVSPNTDEFLDLLASSQSRRLDDQRASFSNLPGLRLTQNSQSVLSHLMTNDNKEADEDFFD
Molecular Weight of Antigen 76.6 kDa
Purification Method Affinity Purified
Quantity 25ul
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 29899
Test Specificity Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.