Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GPT Antibody, Novus Biologicals™
SDP

Catalog No. NBP216731 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP216731 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP216731 Supplier Novus Biologicals Supplier No. NBP216731
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

GPT Polyclonal antibody specifically detects GPT in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin).

Specifications

Antigen GPT
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500-1:3000, Immunohistochemistry 1:100-1:1000, Immunohistochemistry-Paraffin 1:100-1:1000
Formulation 0.1M Tris (pH 7.0), 0.1M Glycine and 10% Glycerol with 0.01% Thimerosal
Gene Accession No. P24298
Gene Alias AAT1alanine aminotransferase 1, ALT1, ALT1EC 2.6.1.2, Glutamate pyruvate transaminase 1, glutamic-alanine transaminase 1, Glutamic--alanine transaminase 1, glutamic-pyruvate transaminase (alanine aminotransferase), Glutamic--pyruvic transaminase 1, GPT1GPT 1
Gene Symbols GPT
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human GPT. The exact sequence is proprietary.
Molecular Weight of Antigen 55 kDa
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline metabolism
Primary or Secondary Primary
Gene ID (Entrez) 2875
Test Specificity Human, Mouse. Expected to cross react based on sequence identity: Rat (88%), Bovine (90%).
Target Species Human, Mouse
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.