Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GPT2 Antibody, PerCP, Novus Biologicals™
SDP

Catalog No. NB292954 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB292954 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB292954 Supplier Novus Biologicals Supplier No. NBP326095PCP
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

GPT2 Polyclonal antibody specifically detects GPT2 in Rat samples. It is validated for ELISA

Specifications

Antigen GPT2
Applications ELISA
Classification Polyclonal
Conjugate PerCP
Dilution ELISA
Formulation PBS
Gene Alias AAT2, alanine aminotransferase 2, ALT2EC 2.6.1.2, Glutamate pyruvate transaminase 2, glutamic pyruvate transaminase (alanine aminotransferase) 2, Glutamic--alanine transaminase 2, glutamic-pyruvate transaminase 2, Glutamic--pyruvic transaminase 2, GPT 2
Host Species Rabbit
Immunogen Rabbits immunized with purified, recombinant Rat GPT2 (Accession#: NP_001012057.1, Met1-Ser522)
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline metabolism
Primary or Secondary Primary
Gene ID (Entrez) 84706
Target Species Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.