Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gr-1/Ly-6G Antibody (1A8), Janelia Fluor™ 646, Novus Biologicals™
SDP

Catalog No. N253131J646 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N253131J646 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N253131J646 Supplier Novus Biologicals Supplier No. NBP253131JF646
Add to Cart
Add to Cart

Rat Monoclonal Antibody

Gr-1/Ly-6G Monoclonal specifically detects Gr-1/Ly-6G in Mouse samples. It is validated for Flow Cytometry.

Specifications

Antigen Gr-1/Ly-6G
Applications Flow Cytometry
Classification Monoclonal
Clone 1A8
Conjugate Janelia Fluor 646
Dilution Flow Cytometry
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias C6orf23, G6D, LY6-D, LY6G6D, lymphocyte antigen 6 family member G6D, MEGT1, NG25
Gene Symbols LY6G6D
Host Species Rat
Immunogen Ly-6G transfected EL-4J cell line.
Purification Method Protein A or G purified
Quantity 0.1 mL
Research Discipline Innate Immunity
Primary or Secondary Primary
Gene ID (Entrez) 58530.0
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.