Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Gr-1/Ly-6G Antibody (8I8), Novus Biologicals™

Human Monoclonal Antibody
$295.74 - $479.73
Specifications
| Antigen | Gr-1/Ly-6G |
|---|---|
| Clone | 8I8 |
| Dilution | Flow Cytometry, ELISA |
| Applications | Flow Cytometry, ELISA |
| Classification | Monoclonal |
Description
Gr-1/Ly-6G Monoclonal antibody specifically detects Gr-1/Ly-6G in Human, Cynomolgus Monkey samples. It is validated for Flow Cytometry, ELISASpecifications
| Gr-1/Ly-6G | |
| Flow Cytometry, ELISA | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Cynomolgus Monkey | |
| C6orf23, G6D, LY6-D, LY6G6D, lymphocyte antigen 6 family member G6D, MEGT1, NG25 | |
| Recombinant Human Gr-1/Ly-6G protein [UniProt O95868] | |
| Primary | |
| Store at -20 to -70C. Avoid freeze-thaw cycles. |
| 8I8 | |
| Flow Cytometry, ELISA | |
| Unconjugated | |
| Human | |
| Innate Immunity | |
| 50% Glycerol, 0.01M PBS, PH 7.4 | |
| 58530 | |
| IgG1 | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title