Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gr-1/Ly-6G Antibody (8I8), Novus Biologicals™
SDP

Catalog No. NB298656 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB298656 50 μL
NB298657 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB298656 Supplier Novus Biologicals Supplier No. NBP32616050UL
Only null left
Add to Cart
Add to Cart

Human Monoclonal Antibody

Gr-1/Ly-6G Monoclonal antibody specifically detects Gr-1/Ly-6G in Human, Cynomolgus Monkey samples. It is validated for Flow Cytometry, ELISA

Specifications

Antigen Gr-1/Ly-6G
Applications Flow Cytometry, ELISA
Classification Monoclonal
Clone 8I8
Conjugate Unconjugated
Dilution Flow Cytometry, ELISA
Formulation 50% Glycerol, 0.01M PBS, PH 7.4
Gene Alias C6orf23, G6D, LY6-D, LY6G6D, lymphocyte antigen 6 family member G6D, MEGT1, NG25
Host Species Human
Immunogen Recombinant Human Gr-1/Ly-6G protein [UniProt O95868]
Purification Method Affinity purified
Quantity 50 μL
Regulatory Status RUO
Research Discipline Innate Immunity
Primary or Secondary Primary
Gene ID (Entrez) 58530
Target Species Human, Cynomolgus Monkey
Content And Storage Store at -20 to -70C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.