Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ GR 125487 sulfamate
GREENER_CHOICESDP

Catalog No. 165810 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
16-581-0 10 mg
16-585-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 16-581-0 Supplier Tocris Bioscience™ Supplier No. 1658/10
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Potent and selective 5-HT4 antagonist; active in vivo

Potent and selective 5-HT4 receptor antagonist (Ki = 0.19 nM for porcine 5-HT4). Centrally active following systemic administration in vivo.

Chemical Identifiers

CAS 859502-43-5
Molecular Formula C19H29FN4O8S2

Specifications

Quantity 10 mg
Formula Weight 524.58
Purity >98%
Molecular Weight (g/mol) 524.579
Chemical Name or Material GR 125487 sulfamate
Recommended Storage Desiccate at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.