Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ GR 127935 hydrochloride
GREENER_CHOICESDP

Catalog No. 147710 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
14-771-0 10 mg
14-775-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 14-771-0 Supplier Tocris Bioscience™ Supplier No. 1477/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Potent and selective 5-HT1B and 5-HT1D antagonist

Potent and selective 5-HT1B/1D receptor antagonist (pKi values are 8.5 for both guinea pig 5-HT1D and rat 5-HT1B receptors). Displays > 100-fold selectivity over 5HT1A, 5-HT2A, 5-HT2C receptors and other receptor types. Centrally active following oral administration.

Chemical Identifiers

CAS 148642-42-6
Molecular Formula C29H32ClN5O3

Specifications

Quantity 10 mg
Formula Weight 534.06
Purity >99%
Molecular Weight (g/mol) 534.057
Chemical Name or Material GR 127935 hydrochloride
Recommended Storage Desiccate at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.