Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ GR 203040
GREENER_CHOICE

Catalog No. p-7139263
Click to view available options
Quantity:
5 mg
25 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Potent, selective NK1 receptor antagonist

GR 203040 is a potent and selective NK1 receptor antagonist. Potently inhibits substance P binding to NK1 receptors (pKi = 10.1 to 10.5). Displays antiemetic activity in vivo.

Specifications

CAS 168398-02-5
Synonym (2S,3S)-N-[[2-Methoxy-5-(1H-tetrazol-1-yl)phenyl]methyl]-2-phenyl-3-piperidinamine dihydrochloride
Quantity 5 mg
Molecular Formula C20H24N6O.2HCl
Formula Weight 437.37
Purity 98%
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.