Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GR/NR3C1 Antibody (BuGR2), Alexa Fluor™ 532, Novus Biologicals™
SDP

Catalog No. NB30731A532 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB30731A532 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB30731A532 Supplier Novus Biologicals Supplier No. NB300731AF532
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

GR/NR3C1 Monoclonal specifically detects GR/NR3C1 in Human, Mouse, Rat, Guinea Pig, Rabbit, Sheep, Yeast samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen GR/NR3C1
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone BuGR2
Conjugate Alexa Fluor 532
Dilution Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias GCR, glucocorticoid receptor, GRGCCR, GRLNuclear receptor subfamily 3 group C member 1, nuclear receptor subfamily 3, group C, member 1, nuclear receptor subfamily 3, group C, member 1 (glucocorticoid receptor)
Gene Symbols NR3C1
Host Species Mouse
Immunogen Partially purified rat GR.
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 2908
Test Specificity Using enzymatic digestion analysis, has been shown to react with the undigested 97 kDa GR, a 17 kDa DNA-binding trypsin fragment, and a 45 kDa steroid- and DNA-binding chymotrypsin fragment.
Target Species Human, Mouse, Rat, Guinea Pig, Rabbit, Sheep, Yeast
Content And Storage Store at 4°C in the dark.
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.