Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GR/NR3C1 Antibody (BuGR2), Allophycocyanin, Novus Biologicals™
SDP

Catalog No. NB300731APC Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB300731APC 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB300731APC Supplier Novus Biologicals Supplier No. NB300731APC
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

GR/NR3C1 Monoclonal specifically detects GR/NR3C1 in Human, Mouse, Rat, Guinea Pig, Rabbit, Sheep, Yeast samples. It is validated for Flow Cytometry.

Specifications

Antigen GR/NR3C1
Applications Flow Cytometry
Classification Monoclonal
Clone BuGR2
Conjugate APC
Dilution Flow Cytometry
Gene Alias GCR, glucocorticoid receptor, GRGCCR, GRLNuclear receptor subfamily 3 group C member 1, nuclear receptor subfamily 3, group C, member 1, nuclear receptor subfamily 3, group C, member 1 (glucocorticoid receptor)
Gene Symbols NR3C1
Host Species Mouse
Immunogen Partially purified rat GR.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Phospho Specific, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 2908
Test Specificity Using enzymatic digestion analysis, has been shown to react with the undigested 97 kDa GR, a 17 kDa DNA-binding trypsin fragment, and a 45 kDa steroid- and DNA-binding chymotrypsin fragment.
Target Species Human, Mouse, Rat, Guinea Pig, Rabbit, Sheep, Yeast
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.